Product Details

SNP ID
rs79130202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76329757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTGGCACGCTTGCTCTGGGGC[G/A]TTCTGGATCTCATAGTAGAAGACGG
Phenotype
MIM: 605356
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
YWHAG PubMed Links

Gene Details

Gene
YWHAG
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012479.3 781 Silent Mutation AAC,AAT N188N NP_036611.2

View Full Product Details