Product Details

SNP ID
rs74449049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:115939818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGATACAATTCTGGTCCATATT[C/G]CTGAGGAAAATATTCAATATGTTCC
Phenotype
MIM: 604732
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TFEC PubMed Links

Gene Details

Gene
TFEC
Gene Name
transcription factor EC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018058.2 2232 UTR 3 NP_001018068.1
NM_001244583.1 2232 UTR 3 NP_001231512.1
NM_012252.3 2232 UTR 3 NP_036384.1
XM_011515963.1 2232 UTR 3 XP_011514265.1
XM_011515964.2 2232 UTR 3 XP_011514266.1
XM_011515965.2 2232 UTR 3 XP_011514267.1
XM_011515966.2 2232 UTR 3 XP_011514268.1
XM_011515969.1 2232 UTR 3 XP_011514271.1
XM_011515970.1 2232 UTR 3 XP_011514272.1
XM_011515972.2 2232 UTR 3 XP_011514274.1
XM_017011874.1 2232 UTR 3 XP_016867363.1
XM_017011875.1 2232 UTR 3 XP_016867364.1
XM_017011876.1 2232 UTR 3 XP_016867365.1
XM_017011877.1 2232 UTR 3 XP_016867366.1
XM_017011878.1 2232 UTR 3 XP_016867367.1

View Full Product Details