Product Details

SNP ID
rs79046186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122698462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTTTTAAGTTACCAATCATAA[A/C]CACAACGACATCTTCAAATGCCTGA
Phenotype
MIM: 609978
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CADPS2 PubMed Links

Gene Details

Gene
CADPS2
Gene Name
calcium dependent secretion activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009571.3 693 Intron NP_001009571.2
NM_001167940.1 693 Intron NP_001161412.1
NM_017954.10 693 Intron NP_060424.9
XM_005250695.4 693 Intron XP_005250752.1
XM_005250696.4 693 Intron XP_005250753.1
XM_005250697.4 693 Intron XP_005250754.1
XM_005250698.4 693 Intron XP_005250755.1
XM_005250699.4 693 Intron XP_005250756.1
XM_005250700.4 693 Intron XP_005250757.1
XM_005250701.4 693 Intron XP_005250758.1
XM_005250702.4 693 Intron XP_005250759.1
XM_005250703.4 693 Intron XP_005250760.1
XM_005250704.4 693 Intron XP_005250761.1
XM_005250705.4 693 Intron XP_005250762.1
XM_005250706.4 693 Intron XP_005250763.1
XM_005250707.4 693 Intron XP_005250764.1
XM_005250708.3 693 Intron XP_005250765.1
XM_011516695.2 693 Intron XP_011514997.1
XM_011516696.2 693 Intron XP_011514998.1
XM_011516697.2 693 Intron XP_011514999.1
XM_017012794.1 693 Intron XP_016868283.1
XM_017012795.1 693 Intron XP_016868284.1
XM_017012796.1 693 Intron XP_016868285.1
XM_017012797.1 693 Intron XP_016868286.1
XM_017012798.1 693 Intron XP_016868287.1
XM_017012799.1 693 Intron XP_016868288.1
XM_017012800.1 693 Intron XP_016868289.1
Gene
RNF133
Gene Name
ring finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139175.1 693 Missense Mutation GTT,TTT V153F NP_631914.1
Gene
RNF148
Gene Name
ring finger protein 148
There are no transcripts associated with this gene.

View Full Product Details