Product Details

SNP ID
rs79582073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128945699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGACTGTGGCAGACTCCCAC[A/G]CTGCAAACCAGGTCTGGGGCTCAGC
Phenotype
MIM: 607218 MIM: 610032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF5 PubMed Links

Gene Details

Gene
IRF5
Gene Name
interferon regulatory factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098627.3 Intron NP_001092097.2
NM_001098629.2 Intron NP_001092099.1
NM_001098630.2 Intron NP_001092100.1
NM_001242452.2 Intron NP_001229381.1
NM_032643.4 Intron NP_116032.1
XM_005250317.3 Intron XP_005250374.1
XM_006715974.2 Intron XP_006716037.1
XM_011516158.2 Intron XP_011514460.1
XM_011516159.2 Intron XP_011514461.1
XM_011516160.1 Intron XP_011514462.1
XM_011516161.1 Intron XP_011514463.1
XM_011516162.1 Intron XP_011514464.1
XM_011516163.2 Intron XP_011514465.1
XM_011516164.1 Intron XP_011514466.1
Gene
TNPO3
Gene Name
transportin 3
There are no transcripts associated with this gene.

View Full Product Details