Product Details

SNP ID
rs75344921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:133257919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGGGTCCAGGCTTTGACCAGG[G/T]CTTTGAACGAACAGCTTTGAGGTTC
Phenotype
MIM: 608185
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXOC4 PubMed Links

Gene Details

Gene
EXOC4
Gene Name
exocyst complex component 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037126.1 Intron NP_001032203.1
NM_021807.3 Intron NP_068579.3
XM_005250523.4 Intron XP_005250580.1
XM_017012494.1 Intron XP_016867983.1
XM_017012495.1 Intron XP_016867984.1

View Full Product Details