Product Details

SNP ID
rs75282408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:129842205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATTCAAATGTTTTGTTGTGTA[A/C]CTAAAAAAAGAAAAGAAAAAAGATC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM49B PubMed Links

Gene Details

Gene
FAM49B
Gene Name
family with sequence similarity 49 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256763.1 1212 Silent Mutation NP_001243692.1
NM_016623.4 1212 Silent Mutation NP_057707.3
XM_005250966.3 1212 Missense Mutation XP_005251023.1
XM_011517108.2 1212 Missense Mutation XP_011515410.1
XM_011517109.2 1212 Missense Mutation XP_011515411.1
XM_011517110.2 1212 Missense Mutation XP_011515412.1
XM_011517111.2 1212 Missense Mutation XP_011515413.1
XM_011517112.2 1212 Missense Mutation XP_011515414.1
XM_011517113.2 1212 Missense Mutation XP_011515415.1
XM_011517114.2 1212 Missense Mutation XP_011515416.1
XM_011517115.2 1212 Missense Mutation XP_011515417.1
XM_011517116.2 1212 Missense Mutation XP_011515418.1
XM_011517117.2 1212 Missense Mutation XP_011515419.1
XM_011517118.2 1212 Missense Mutation XP_011515420.1
XM_011517120.2 1212 Missense Mutation XP_011515422.1
XM_011517121.2 1212 Missense Mutation XP_011515423.1
XM_011517122.2 1212 Missense Mutation XP_011515424.1
XM_011517123.2 1212 Missense Mutation XP_011515425.1
XM_011517124.2 1212 Missense Mutation XP_011515426.1
XM_011517125.2 1212 Missense Mutation XP_011515427.1
XM_017013539.1 1212 Missense Mutation XP_016869028.1
XM_017013540.1 1212 Missense Mutation XP_016869029.1
XM_017013541.1 1212 Missense Mutation XP_016869030.1
XM_017013542.1 1212 Missense Mutation XP_016869031.1
XM_017013543.1 1212 Missense Mutation XP_016869032.1
XM_017013544.1 1212 Missense Mutation XP_016869033.1
XM_017013545.1 1212 Missense Mutation XP_016869034.1
XM_017013546.1 1212 Missense Mutation XP_016869035.1
XM_017013547.1 1212 Missense Mutation XP_016869036.1
XM_017013548.1 1212 Missense Mutation XP_016869037.1
XM_017013549.1 1212 Missense Mutation XP_016869038.1
XM_017013550.1 1212 Missense Mutation XP_016869039.1
XM_017013551.1 1212 Missense Mutation XP_016869040.1
XM_017013552.1 1212 Missense Mutation XP_016869041.1
XM_017013553.1 1212 Missense Mutation XP_016869042.1
XM_017013554.1 1212 Missense Mutation XP_016869043.1
XM_017013555.1 1212 Missense Mutation XP_016869044.1
XM_017013556.1 1212 Missense Mutation XP_016869045.1
XM_017013557.1 1212 Missense Mutation XP_016869046.1
XM_017013558.1 1212 Missense Mutation XP_016869047.1
XM_017013559.1 1212 Missense Mutation XP_016869048.1
XM_017013560.1 1212 Silent Mutation XP_016869049.1
XM_017013561.1 1212 Missense Mutation XP_016869050.1
XM_017013562.1 1212 Missense Mutation XP_016869051.1

View Full Product Details