Product Details
- SNP ID
-
rs78880232
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:134600623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATACAGATGTTCCCGGGTGTGG[G/T]TGCGGACATGCATATCAAAATGCAC
- Phenotype
-
MIM: 610931
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZFAT
PubMed Links
Gene Details
- Gene
- ZFAT
- Gene Name
- zinc finger and AT-hook domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001029939.3 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
NP_001025110.2 |
NM_001167583.2 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
NP_001161055.1 |
NM_001174157.1 |
2633 |
Missense Mutation |
AAC,ACC |
N701T |
NP_001167628.1 |
NM_001174158.1 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
NP_001167629.1 |
NM_001289394.1 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
NP_001276323.1 |
NM_020863.3 |
2633 |
Missense Mutation |
AAC,ACC |
N763T |
NP_065914.2 |
XM_011517203.1 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
XP_011515505.1 |
XM_011517204.2 |
2633 |
Missense Mutation |
AAC,ACC |
N701T |
XP_011515506.1 |
XM_011517206.1 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
XP_011515508.1 |
XM_017013716.1 |
2633 |
Missense Mutation |
AAC,ACC |
N751T |
XP_016869205.1 |
- Gene
- ZFAT-AS1
- Gene Name
- ZFAT antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details