Product Details
- SNP ID
-
rs76379783
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143247622 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATATTACAAGTGAAGGGTCCTCC[G/T]CCCAGAGAGGCGACGGAGCTACGCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZFP41
PubMed Links
Gene Details
- Gene
- ZFP41
- Gene Name
- ZFP41 zinc finger protein
View Full Product Details