Product Details

SNP ID
rs76896296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19508027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCGTATCTGAAGAACAAAAGTA[C/T]AATTTCTCTTGCTTTTAGTGCTAGC
Phenotype
MIM: 609838
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC24A2 PubMed Links

Gene Details

Gene
SLC24A2
Gene Name
solute carrier family 24 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193288.2 10214 UTR 3 NP_001180217.1
NM_020344.3 10214 UTR 3 NP_065077.1
XM_005251425.2 10214 UTR 3 XP_005251482.1
XM_005251426.4 10214 UTR 3 XP_005251483.1
XM_006716750.3 10214 UTR 3 XP_006716813.1
XM_017014592.1 10214 UTR 3 XP_016870081.1
XM_017014593.1 10214 UTR 3 XP_016870082.1

View Full Product Details