Product Details
- SNP ID
-
rs79225611
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21077384 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCACTCTGACTATGGTCCAGGC[A/G]CAGTGACTGTACTCCTTGGCCTTCA
- Phenotype
-
MIM: 147640
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNB1
PubMed Links
Gene Details
- Gene
- IFNB1
- Gene Name
- interferon beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002176.3 |
580 |
Silent Mutation |
TGC,TGT |
C162C |
NP_002167.1 |
View Full Product Details