Product Details

SNP ID
rs80275122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34338777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCAGCCCTGTTTTACAGGGAG[C/T]CCTGGAGGAGTTGGGATAGAGGCCA
Phenotype
MIM: 613747 MIM: 602852
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
There are no transcripts associated with this gene.

Gene
NUDT2
Gene Name
nudix hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161.4 159 UTR 5 NP_001152.1
NM_001244390.1 159 Intron NP_001231319.1
NM_147172.2 159 UTR 5 NP_671701.1
NM_147173.2 159 UTR 5 NP_671702.1

View Full Product Details