Product Details
- SNP ID
-
rs76374020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:71866930 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACGTTCAAACAATGCGAGGCCAT[C/G]TGAAAAGTCAATGACTTCATCTTCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD17B
PubMed Links
Gene Details
- Gene
- ABHD17B
- Gene Name
- abhydrolase domain containing 17B
There are no transcripts associated with this gene.
View Full Product Details