Product Details

SNP ID
rs78219814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16440958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGATACCTTGGGAGCTGTGGGG[A/C]GGGCCCTGAAAGAGGCTTGTCCTAT
Phenotype
MIM: 604446
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTER PubMed Links

Gene Details

Gene
PTER
Gene Name
phosphotriesterase related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001484.2 Intron NP_001001484.1
NM_001261836.1 Intron NP_001248765.1
NM_001261837.1 Intron NP_001248766.1
NM_001261838.1 Intron NP_001248767.1
NM_030664.4 Intron NP_109589.2
XM_017016927.1 Intron XP_016872416.1
XM_017016928.1 Intron XP_016872417.1
XM_017016929.1 Intron XP_016872418.1
XM_017016930.1 Intron XP_016872419.1

View Full Product Details