Product Details

SNP ID
rs76052898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5393860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCGATTGCTGCATTCACTTCC[C/T]TGGGGACCACATCCCCTCTATAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBAL3 PubMed Links

Gene Details

Gene
TUBAL3
Gene Name
tubulin alpha like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171864.1 1036 Missense Mutation AAG,AGG K293R NP_001165335.1
NM_024803.2 1036 Missense Mutation AAG,AGG K333R NP_079079.1

View Full Product Details