Product Details

SNP ID
rs78465464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:521966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAAACAGCTCTCATAACCCAAA[C/T]GAAGGCAGAAAAGGGAAGATAAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101059906 PubMed Links

Gene Details

Gene
LOC101059906
Gene Name
collagen alpha-2(IV) chain-like
There are no transcripts associated with this gene.

Gene
LRRC56
Gene Name
leucine rich repeat containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198075.3 Intron NP_932341.1
XM_005252775.2 Intron XP_005252832.1
XM_005252776.2 Intron XP_005252833.1
XM_006718132.2 Intron XP_006718195.1
XM_006718133.2 Intron XP_006718196.1
XM_011519875.2 Intron XP_011518177.1
XM_011519876.1 Intron XP_011518178.1
XM_011519877.2 Intron XP_011518179.1
XM_017017167.1 Intron XP_016872656.1
XM_017017168.1 Intron XP_016872657.1

View Full Product Details