Product Details

SNP ID
rs75694066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46632127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCTATAAGTATCAAGTCACAG[A/G]ACAATAAGTGTAGATGAAGTTTAAA
Phenotype
MIM: 615088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG13 PubMed Links

Gene Details

Gene
ATG13
Gene Name
autophagy related 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142673.2 Intron NP_001136145.1
NM_001205119.1 Intron NP_001192048.1
NM_001205120.1 Intron NP_001192049.1
NM_001205121.1 Intron NP_001192050.1
NM_001205122.1 Intron NP_001192051.1
NM_014741.4 Intron NP_055556.2
XM_005253262.2 Intron XP_005253319.1
XM_005253263.2 Intron XP_005253320.1
XM_005253265.2 Intron XP_005253322.1
XM_005253266.2 Intron XP_005253323.1
XM_005253268.2 Intron XP_005253325.1
XM_006718394.2 Intron XP_006718457.1
XM_006718395.2 Intron XP_006718458.1
XM_006718396.2 Intron XP_006718459.1
XM_011520493.1 Intron XP_011518795.1
XM_011520496.2 Intron XP_011518798.1
XM_011520499.2 Intron XP_011518801.1
XM_011520500.1 Intron XP_011518802.1
XM_011520501.1 Intron XP_011518803.1
XM_011520502.1 Intron XP_011518804.1
XM_011520503.1 Intron XP_011518805.1
XM_017018599.1 Intron XP_016874088.1
XM_017018600.1 Intron XP_016874089.1
XM_017018601.1 Intron XP_016874090.1
XM_017018602.1 Intron XP_016874091.1
XM_017018603.1 Intron XP_016874092.1
XM_017018604.1 Intron XP_016874093.1
XM_017018605.1 Intron XP_016874094.1
XM_017018606.1 Intron XP_016874095.1
XM_017018607.1 Intron XP_016874096.1
XM_017018608.1 Intron XP_016874097.1
XM_017018609.1 Intron XP_016874098.1
XM_017018610.1 Intron XP_016874099.1
XM_017018611.1 Intron XP_016874100.1
XM_017018612.1 Intron XP_016874101.1
XM_017018613.1 Intron XP_016874102.1
XM_017018614.1 Intron XP_016874103.1
XM_017018615.1 Intron XP_016874104.1
XM_017018616.1 Intron XP_016874105.1
XM_017018617.1 Intron XP_016874106.1
XM_017018618.1 Intron XP_016874107.1
XM_017018619.1 Intron XP_016874108.1
XM_017018620.1 Intron XP_016874109.1
XM_017018621.1 Intron XP_016874110.1
XM_017018622.1 Intron XP_016874111.1

View Full Product Details