Product Details

SNP ID
rs77633179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8982313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCTGCTCTGCCTCTTTCCTT[A/G]TATCAGGACAGAACTGAACATCTCA
Phenotype
MIM: 613125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRIP3 PubMed Links

Gene Details

Gene
NRIP3
Gene Name
nuclear receptor interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020645.2 2072 UTR 3 NP_065696.1
Gene
TMEM9B-AS1
Gene Name
TMEM9B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details