Product Details

SNP ID
rs78571415
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:32687161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAGGCTAATTTATCTTTTTTTT[C/T]CCTTTTATTGCTTTATGGTGTCATA
Phenotype
MIM: 603850
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DNM1L PubMed Links
Additional Information
For this assay, SNP(s) [rs151269726] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DNM1L
Gene Name
dynamin 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278463.1 Intron NP_001265392.1
NM_001278464.1 Intron NP_001265393.1
NM_001278465.1 Intron NP_001265394.1
NM_001278466.1 Intron NP_001265395.1
NM_005690.4 Intron NP_005681.2
NM_012062.4 Intron NP_036192.2
NM_012063.3 Intron NP_036193.2
XM_005253282.4 Intron XP_005253339.1
XM_011520543.2 Intron XP_011518845.1
XM_011520544.2 Intron XP_011518846.1
XM_017018663.1 Intron XP_016874152.1
XM_017018664.1 Intron XP_016874153.1
XM_017018665.1 Intron XP_016874154.1

View Full Product Details