Product Details

SNP ID
rs75688283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10158918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCCTAAAAATAGAAGATATAT[G/T]TCTAATTCCATCTGTTTCTATTCAG
Phenotype
MIM: 602601
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172632.1 1758 UTR 3 NP_001166103.1
NM_001172633.1 1758 UTR 3 NP_001166104.1
NM_002543.3 1758 UTR 3 NP_002534.1

View Full Product Details