Product Details

SNP ID
rs77049450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12476412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACAGGGTCTACCTACCTGACCC[C/G]CAAGCTGGCTCAGTCTCAGCGCTAA
Phenotype
MIM: 616598 MIM: 607175
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BORCS5 PubMed Links

Gene Details

Gene
BORCS5
Gene Name
BLOC-1 related complex subunit 5
There are no transcripts associated with this gene.

Gene
DUSP16
Gene Name
dual specificity phosphatase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030640.2 2910 UTR 3 NP_085143.1
XM_005253488.2 2910 UTR 3 XP_005253545.1
XM_006719155.2 2910 UTR 3 XP_006719218.1
XM_011520856.1 2910 UTR 3 XP_011519158.1
XM_011520857.1 2910 UTR 3 XP_011519159.1
XM_017019988.1 2910 UTR 3 XP_016875477.1
Gene
LOC105369662
Gene Name
uncharacterized LOC105369662
There are no transcripts associated with this gene.

View Full Product Details