Product Details

SNP ID
rs74344050
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:129073992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTACTCATCACGATGGAGTTCCT[A/G]GTGGGGTACTCGTCGTCTGAGGAGA
Phenotype
MIM: 611257
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMEM132D PubMed Links

Gene Details

Gene
TMEM132D
Gene Name
transmembrane protein 132D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133448.2 3511 Silent Mutation NP_597705.2

View Full Product Details