Product Details

SNP ID
rs79948938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121778624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTTCTCAAAGGAGGCCAGGAT[A/G]TCTCGCCTCCTGAACTCCACAGGCT
Phenotype
MIM: 616551
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHOF PubMed Links

Gene Details

Gene
RHOF
Gene Name
ras homolog family member F, filopodia associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019034.2 4066 UTR 3 NP_061907.2
Gene
TMEM120B
Gene Name
transmembrane protein 120B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080825.2 4066 UTR 3 NP_001074294.2
XM_005253843.4 4066 Intron XP_005253900.1
XM_017018853.1 4066 Intron XP_016874342.1
XM_017018854.1 4066 Intron XP_016874343.1

View Full Product Details