Product Details

SNP ID
rs78107102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110502080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCGGGCTCCGCTCAGTCACCA[A/C]CACCGTCGCCGCCCTCTTCCTCAGG
Phenotype
MIM: 608368 MIM: 606932
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAD9B PubMed Links

Gene Details

Gene
RAD9B
Gene Name
RAD9 checkpoint clamp component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286531.1 30 Intron NP_001273460.1
NM_001286532.1 30 Intron NP_001273461.1
NM_001286533.1 30 Intron NP_001273462.1
NM_001286534.1 30 Intron NP_001273463.1
NM_001286535.1 30 Intron NP_001273464.1
NM_001286536.1 30 Intron NP_001273465.1
NM_152442.3 30 Intron NP_689655.3
XM_005253849.4 30 Intron XP_005253906.1
XM_011537971.2 30 Intron XP_011536273.1
XM_011537972.2 30 Intron XP_011536274.1
XM_011537974.2 30 Intron XP_011536276.1
XM_017018877.1 30 Intron XP_016874366.1
XM_017018878.1 30 Intron XP_016874367.1
XM_017018879.1 30 Intron XP_016874368.1
XM_017018880.1 30 Intron XP_016874369.1
XM_017018881.1 30 Intron XP_016874370.1
XM_017018882.1 30 Intron XP_016874371.1
Gene
VPS29
Gene Name
VPS29, retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282150.1 30 UTR 5 NP_001269079.1
NM_001282151.1 30 UTR 5 NP_001269080.1
NM_016226.4 30 UTR 5 NP_057310.1
NM_057180.2 30 UTR 5 NP_476528.1
XM_006719459.3 30 UTR 5 XP_006719522.1
XM_006719460.3 30 UTR 5 XP_006719523.1
XM_011538484.2 30 UTR 5 XP_011536786.1
XM_017019457.1 30 Intron XP_016874946.1
XM_017019458.1 30 Intron XP_016874947.1
XM_017019459.1 30 Intron XP_016874948.1

View Full Product Details