Product Details

SNP ID
rs76879767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116718234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAAACAGTCTAAGATCCAATGG[A/G]TGTTTTGCACCTGATCCTGAGCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf49 PubMed Links

Gene Details

Gene
C12orf49
Gene Name
chromosome 12 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024738.2 Intron NP_079014.1
XM_005253937.3 Intron XP_005253994.1

View Full Product Details