Product Details

SNP ID
rs75177904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:97447585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGAGACCCCTTATGCATCAAAT[C/T]AATTTAAGCATTTCAGTGTCTAACA
Phenotype
MIM: 179540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAP2A PubMed Links

Gene Details

Gene
RAP2A
Gene Name
RAP2A, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021033.6 Intron NP_066361.1

View Full Product Details