Product Details

SNP ID
rs79336396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40895864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAAGTGGCGAGGAGCACTGTG[G/T]TGTTTTTTCTGCCTCTCCTTGCCCT
Phenotype
MIM: 608551
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS18 PubMed Links

Gene Details

Gene
VPS18
Gene Name
VPS18, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020857.2 Intron NP_065908.1
XM_011521843.2 Intron XP_011520145.1
XM_011521844.2 Intron XP_011520146.1
XM_017022447.1 Intron XP_016877936.1

View Full Product Details