Product Details

SNP ID
rs76674534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50908961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCGGCCCGGGAGCTCAGGGAC[A/G]TCGGAGTGAGGCCCCCGCGCCAGGA
Phenotype
MIM: 607244
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP4E1 PubMed Links

Gene Details

Gene
AP4E1
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252127.1 Intron NP_001239056.1
NM_007347.4 Intron NP_031373.2
XM_005254264.3 Intron XP_005254321.1
XM_006720447.3 Intron XP_006720510.1
XM_017022042.1 Intron XP_016877531.1

View Full Product Details