Product Details

SNP ID
rs77996726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:31327239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGCCGCCGCCTATGTGGACCA[C/T]TTCGCCGCCGAGTGCCTCGTGTCCA
Phenotype
MIM: 605328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF13 PubMed Links

Gene Details

Gene
KLF13
Gene Name
Kruppel like factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302461.1 385 Silent Mutation CAC,CAT H9H NP_001289390.1
NM_015995.3 385 Silent Mutation CAC,CAT H9H NP_057079.2

View Full Product Details