Product Details

SNP ID
rs78920779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66550373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTAAAAGTAGCAAATGCCTTAT[A/G]GTTTTTAGTCACTGCTTCCATTTAA
Phenotype
MIM: 609984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCTL PubMed Links

Gene Details

Gene
LCTL
Gene Name
lactase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278562.1 Intron NP_001265491.1
NM_207338.3 Intron NP_997221.2
XM_017021999.1 Intron XP_016877488.1
Gene
ZWILCH
Gene Name
zwilch kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287821.1 Intron NP_001274750.1
NM_001287822.1 Intron NP_001274751.1
NM_001287823.1 Intron NP_001274752.1
NM_017975.4 Intron NP_060445.3

View Full Product Details