Product Details

SNP ID
rs75552790
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75906314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTAGACTCTCTAAGTGTACTC[C/T]TTGCCATTGAGGGATTTGCTGCTCC
Phenotype
MIM: 609096 MIM: 612501
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXO22 PubMed Links

Gene Details

Gene
FBXO22
Gene Name
F-box protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012170.3 Intron NP_036302.1
NM_147188.2 Intron NP_671717.1
XM_011521447.2 Intron XP_011519749.1
Gene
UBE2Q2
Gene Name
ubiquitin conjugating enzyme E2 Q2
There are no transcripts associated with this gene.

View Full Product Details