Product Details
- SNP ID
-
rs76788604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78090639 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGCATCACCATCATCATCATCAT[A/C]ATCATCATCATCATCATCATCATCA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149544016] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SH2D7
- Gene Name
- SH2 domain containing 7
View Full Product Details