Product Details
- SNP ID
-
rs76961090
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:38455477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGTTGAACCCTGATAATACCTAC[C/T]TTGCAGTGACCCAGGGTTACGGAGT
- Phenotype
-
MIM: 616142
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM98B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138505347] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM98B
- Gene Name
- family with sequence similarity 98 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173611.3 |
|
Intron |
|
|
NP_775882.2 |
View Full Product Details