Product Details

SNP ID
rs75618070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88812824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGAGCTCCACAAAGAGCACT[C/T]CTTCTCACTGCACCAAGGGGAGTGA
Phenotype
MIM: 155760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAN PubMed Links

Gene Details

Gene
ACAN
Gene Name
aggrecan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135.3 Intron NP_001126.3
NM_013227.3 Intron NP_037359.3
XM_006720419.1 Intron XP_006720482.1
XM_011521313.1 Intron XP_011519615.1
XM_011521314.1 Intron XP_011519616.1
XM_017021985.1 Intron XP_016877474.1
XM_017021986.1 Intron XP_016877475.1
XM_017021987.1 Intron XP_016877476.1

View Full Product Details