Product Details

SNP ID
rs78194647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90931249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGCCTGAACGAGGCTGGAGTTG[G/T]GCCTCTGGATAGCTTCAAGCACTGA
Phenotype
MIM: 600988 MIM: 611219
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HDDC3 PubMed Links

Gene Details

Gene
HDDC3
Gene Name
HD domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286451.1 595 UTR 3 NP_001273380.1
NM_198527.3 595 UTR 3 NP_940929.1
Gene
MAN2A2
Gene Name
mannosidase alpha class 2A member 2
There are no transcripts associated with this gene.

Gene
UNC45A
Gene Name
unc-45 myosin chaperone A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039675.1 595 Intron NP_001034764.1
NM_001323619.1 595 Intron NP_001310548.1
NM_001323620.1 595 Intron NP_001310549.1
NM_001323621.1 595 Intron NP_001310550.1
NM_018671.4 595 Intron NP_061141.2
XM_005254963.3 595 Intron XP_005255020.1
XM_011521779.2 595 Intron XP_011520081.1
XM_011521780.2 595 Intron XP_011520082.1
XM_017022407.1 595 Intron XP_016877896.1

View Full Product Details