Product Details

SNP ID
rs75954521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:91101377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTGTTCTGCTTGTTCCAGTTGG[C/T]GCAGTAGGTACAGGCTCGTTCTGCA
Phenotype
MIM: 185861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SV2B PubMed Links

Gene Details

Gene
SV2B
Gene Name
synaptic vesicle glycoprotein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167580.1 Intron NP_001161052.1
NM_001323031.1 Intron NP_001309960.1
NM_001323032.1 Intron NP_001309961.1
NM_001323033.1 Intron NP_001309962.1
NM_001323034.1 Intron NP_001309963.1
NM_001323036.1 Intron NP_001309965.1
NM_001323037.1 Intron NP_001309966.1
NM_001323038.1 Intron NP_001309967.1
NM_001323039.1 Intron NP_001309968.1
NM_001323040.1 Intron NP_001309969.1
NM_014848.5 Intron NP_055663.1
XM_005254998.3 Intron XP_005255055.1
XM_017022761.1 Intron XP_016878250.1
XM_017022762.1 Intron XP_016878251.1

View Full Product Details