Product Details

SNP ID
rs77983756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27270252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTAATTTAAAGATGATTTTCAA[C/T]ATACTGGAGGATTTGTGTAGGTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ21408 PubMed Links

Gene Details

Gene
FLJ21408
Gene Name
uncharacterized LOC400512
There are no transcripts associated with this gene.

Gene
NSMCE1
Gene Name
NSE1 homolog, SMC5-SMC6 complex component
There are no transcripts associated with this gene.

View Full Product Details