Product Details

SNP ID
rs76148227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2895170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGCAGCACCAGTGTGGGGTGG[C/T]CCGGCACTTGCCCCTACAGGGTTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLYWCH2 PubMed Links

Gene Details

Gene
FLYWCH2
Gene Name
FLYWCH family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142499.1 Intron NP_001135971.1
NM_001142500.1 Intron NP_001135972.1
NM_138439.2 Intron NP_612448.1
XM_005255078.4 Intron XP_005255135.1

View Full Product Details