Product Details

SNP ID
rs79568162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29813493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCCCAACCAGACCCCCGGCCA[C/G]ATTCCCAGCCTACCCCCAAGCCAGC
Phenotype
MIM: 603213 MIM: 600999 MIM: 605088 MIM: 612033 MIM: 614386
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
MAZ
Gene Name
MYC associated zinc finger protein
There are no transcripts associated with this gene.

Gene
MVP
Gene Name
major vault protein
There are no transcripts associated with this gene.

Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256442.1 735 Missense Mutation CAT,GAT H147D NP_001243371.1
NM_001256443.1 735 Missense Mutation CAT,GAT H147D NP_001243372.1
NM_145239.2 735 Missense Mutation CAT,GAT H147D NP_660282.2
XM_011545715.2 735 Missense Mutation CAT,GAT H147D XP_011544017.1
XM_011545716.2 735 Missense Mutation CAT,GAT H147D XP_011544018.1
XM_017022887.1 735 Missense Mutation CAT,GAT H147D XP_016878376.1
XM_017022888.1 735 Missense Mutation CAT,GAT H147D XP_016878377.1
XM_017022889.1 735 Missense Mutation CAT,GAT H147D XP_016878378.1

View Full Product Details