Product Details

SNP ID
rs74781964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30417983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCTTTGGCGGCAGCTCGTGCCT[C/G]GCGCGCCACCGACGCACGCACACGG
Phenotype
MIM: 615840
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCTPP1 PubMed Links

Gene Details

Gene
DCTPP1
Gene Name
dCTP pyrophosphatase 1
There are no transcripts associated with this gene.

Gene
ZNF771
Gene Name
zinc finger protein 771
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142305.1 843 Silent Mutation CTC,CTG L190L NP_001135777.1
NM_016643.3 843 Silent Mutation CTC,CTG L190L NP_057727.2

View Full Product Details