Product Details

SNP ID
rs74973579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30752684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCCTCTGCAAAAGTCAGAGGG[A/G]ACGTAAGGATGTCTATTATGCCATC
Phenotype
MIM: 172471 MIM: 607700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC189 PubMed Links

Gene Details

Gene
CCDC189
Gene Name
coiled-coil domain containing 189
There are no transcripts associated with this gene.

Gene
PHKG2
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000294.2 Intron NP_000285.1
NM_001172432.1 Intron NP_001165903.1
Gene
RNF40
Gene Name
ring finger protein 40
There are no transcripts associated with this gene.

Gene
TMEM265
Gene Name
transmembrane protein 265
There are no transcripts associated with this gene.

View Full Product Details