Product Details

SNP ID
rs74795295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56445615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCAGTTTTCAGCCCATGCATG[C/T]CCCCACTTTCATAAGTACCTAGCAC
Phenotype
MIM: 604978 MIM: 615857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 Intron NP_008937.1
Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details