Product Details

SNP ID
rs79668208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84976339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAAGTTTCAGTCTGATGAGCCA[C/T]GCCTCACACTGAGAACTCCGGGCCA
Phenotype
MIM: 614604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC7 PubMed Links

Gene Details

Gene
ZDHHC7
Gene Name
zinc finger DHHC-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145548.1 795 UTR 3 NP_001139020.1
NM_017740.2 795 UTR 3 NP_060210.2
XM_011523222.1 795 Intron XP_011521524.1
XM_017023434.1 795 UTR 3 XP_016878923.1
XM_017023435.1 795 UTR 3 XP_016878924.1
XM_017023436.1 795 UTR 3 XP_016878925.1

View Full Product Details