Product Details

SNP ID
rs78686908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53057813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAAGGGCAAACAGGAGGACAC[A/G]CTGCTTTCTGGTTTCAGCTCTTGTG
Phenotype
MIM: 616936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHD9 PubMed Links

Gene Details

Gene
CHD9
Gene Name
chromodomain helicase DNA binding protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308319.1 Intron NP_001295248.1
NM_025134.4 Intron NP_079410.4
XM_005256168.3 Intron XP_005256225.1
XM_005256169.2 Intron XP_005256226.1
XM_005256172.3 Intron XP_005256229.1
XM_005256176.3 Intron XP_005256233.1
XM_006721281.2 Intron XP_006721344.1
XM_006721283.3 Intron XP_006721346.1
XM_011523347.2 Intron XP_011521649.1
XM_017023718.1 Intron XP_016879207.1
XM_017023719.1 Intron XP_016879208.1
XM_017023720.1 Intron XP_016879209.1
XM_017023721.1 Intron XP_016879210.1
XM_017023722.1 Intron XP_016879211.1
XM_017023723.1 Intron XP_016879212.1
XM_017023724.1 Intron XP_016879213.1
XM_017023725.1 Intron XP_016879214.1
XM_017023726.1 Intron XP_016879215.1
XM_017023727.1 Intron XP_016879216.1
XM_017023728.1 Intron XP_016879217.1
Gene
LOC105371267
Gene Name
p53-regulated lncRNA 1
There are no transcripts associated with this gene.

View Full Product Details