Product Details

SNP ID
rs76267236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88864866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGGCGCACCTTGATGACCCGG[C/T]CCCGGAAGAGGCTCTGGTCCAGCTC
Phenotype
MIM: 603870 MIM: 612222 MIM: 610970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBFA2T3 PubMed Links

Gene Details

Gene
CBFA2T3
Gene Name
CBFA2/RUNX1 translocation partner 3
There are no transcripts associated with this gene.

Gene
GALNS
Gene Name
galactosamine (N-acetyl)-6-sulfatase
There are no transcripts associated with this gene.

Gene
PABPN1L
Gene Name
poly(A) binding protein nuclear 1 like (cytoplasmic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080487.2 1950 Missense Mutation GAC,GGC D214G NP_001073956.2
NM_001294328.1 1950 Intron NP_001281257.1
XM_017023230.1 1950 Missense Mutation GAC,GGC D214G XP_016878719.1
Gene
TRAPPC2L
Gene Name
trafficking protein particle complex 2-like
There are no transcripts associated with this gene.

View Full Product Details