Product Details

SNP ID
rs75884483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16622219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTCCTAATGAAGATTTTCCTA[C/T]ATTCATAAAATATAGTTTATAAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF624 PubMed Links

Gene Details

Gene
ZNF624
Gene Name
zinc finger protein 624
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020787.3 2759 UTR 3 NP_065838.2
XM_006721562.3 2759 UTR 3 XP_006721625.1
XM_011523970.2 2759 UTR 3 XP_011522272.1

View Full Product Details