Product Details

SNP ID
rs74349194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5183423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATGTGGGACAAGGTGTGAACT[C/T]TGACTGAACATCTGTCCACAGTCAT
Phenotype
MIM: 604334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130950 PubMed Links

Gene Details

Gene
LOC100130950
Gene Name
uncharacterized LOC100130950
There are no transcripts associated with this gene.

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
There are no transcripts associated with this gene.

Gene
ZNF594
Gene Name
zinc finger protein 594
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032530.1 1303 Silent Mutation CAA,CAG Q278Q NP_115919.1
XM_005256827.2 1303 Silent Mutation CAA,CAG Q278Q XP_005256884.1

View Full Product Details