Product Details

SNP ID
rs78359780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68273878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCCTCCTTCCCCTTCCTTGTT[C/T]TTCAAACTAAGTGTCTAGACAACTT
Phenotype
MIM: 610008 MIM: 603880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSG PubMed Links

Gene Details

Gene
ARSG
Gene Name
arylsulfatase G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267727.1 Intron NP_001254656.1
NM_014960.4 Intron NP_055775.2
XM_005257170.3 Intron XP_005257227.1
XM_005257172.3 Intron XP_005257229.1
XM_006721777.3 Intron XP_006721840.2
XM_006721779.3 Intron XP_006721842.1
XM_011524535.2 Intron XP_011522837.1
XM_011524536.2 Intron XP_011522838.1
XM_011524537.1 Intron XP_011522839.1
XM_011524538.2 Intron XP_011522840.1
XM_011524540.2 Intron XP_011522842.1
XM_011524541.2 Intron XP_011522843.1
XM_011524542.2 Intron XP_011522844.1
XM_011524543.2 Intron XP_011522845.1
XM_011524544.2 Intron XP_011522846.1
XM_011524545.2 Intron XP_011522847.1
XM_011524546.2 Intron XP_011522848.1
XM_017024360.1 Intron XP_016879849.1
XM_017024361.1 Intron XP_016879850.1
XM_017024362.1 Intron XP_016879851.1
XM_017024363.1 Intron XP_016879852.1
XM_017024364.1 Intron XP_016879853.1
XM_017024365.1 Intron XP_016879854.1
XM_017024366.1 Intron XP_016879855.1
XM_017024367.1 Intron XP_016879856.1
XM_017024368.1 Intron XP_016879857.1
Gene
SLC16A6
Gene Name
solute carrier family 16 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174166.1 Intron NP_001167637.1
NM_004694.4 Intron NP_004685.2
XM_005257789.3 Intron XP_005257846.2
XM_011525461.2 Intron XP_011523763.1
XM_017025291.1 Intron XP_016880780.1
XM_017025292.1 Intron XP_016880781.1

View Full Product Details