Product Details

SNP ID
rs80341494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79834812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCACCTCGCCGGACTTGATCTTC[A/C]CCGCCTGCATGCCGTTCTCCATGTA
Phenotype
MIM: 603079
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CBX4 PubMed Links

Gene Details

Gene
CBX4
Gene Name
chromobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003655.2 2502 Missense Mutation GGG,GTG G277V NP_003646.2
XM_011525399.2 2502 Missense Mutation GGG,GTG G211V XP_011523701.1

View Full Product Details