Product Details

SNP ID
rs78234209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28723781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTTGACAAACCTTATCCTCAC[A/G]TTCCTCATTTTGCTTTCTAAAAATA
Phenotype
MIM: 609799 MIM: 610917 MIM: 602326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NARR PubMed Links

Gene Details

Gene
NARR
Gene Name
nine-amino acid residue-repeats
There are no transcripts associated with this gene.

Gene
NEK8
Gene Name
NIMA related kinase 8
There are no transcripts associated with this gene.

Gene
RAB34
Gene Name
RAB34, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RPL23A
Gene Name
ribosomal protein L23a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000984.5 Intron NP_000975.2
Gene
SNORD42A
Gene Name
small nucleolar RNA, C/D box 42A
There are no transcripts associated with this gene.

Gene
SNORD42B
Gene Name
small nucleolar RNA, C/D box 42B
There are no transcripts associated with this gene.

Gene
SNORD4A
Gene Name
small nucleolar RNA, C/D box 4A
There are no transcripts associated with this gene.

Gene
SNORD4B
Gene Name
small nucleolar RNA, C/D box 4B
There are no transcripts associated with this gene.

Gene
TLCD1
Gene Name
TLC domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160407.1 Intron NP_001153879.1
NM_138463.3 Intron NP_612472.1
XM_006721671.3 Intron XP_006721734.1
XM_011524278.2 Intron XP_011522580.1

View Full Product Details