Product Details
- SNP ID
-
rs77898691
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51317834 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCCTTGCAGGGAAGAACTACTAT[C/T]TTTCACCTTGTACAGTTGGAGAAAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IGLON5
PubMed Links
Gene Details
- Gene
- IGLON5
- Gene Name
- IgLON family member 5
View Full Product Details